Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kir5.1 (KCNJ16) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541051
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541051 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541051 Supplier Invitrogen™ Supplier No. PA541051
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RESCTSDTKA RRRSFSAVAI VSSCENPEET TTSATHEYRE TPYQKALLTL Sequence homology: Dog: 86%; Horse: 93%; Human: 100%; Pig: 79%; Rabbit: 86%.

Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. KCNJ16 is an integral membrane protein and inward-rectifier type potassium channel. KCNJ16, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, can form heterodimers with two other inward-rectifier type potassium channels. It may be involved in the regulation of fluid and pH balance.Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. The encoded protein, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, can form heterodimers with two other inward-rectifier type potassium channels. It may be involved in the regulation of fluid and pH balance. Three transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kir5.1 (KCNJ16)
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Kcnj16
Gene Accession No. Q9NPI9
Gene Alias 6430410F18Rik; AI132396; BIR9; Inward rectifier K(+) channel Kir5.1; inward rectifier K+ channel KIR5.1; inward rectifier potassium channel 16; inwardly-rectifying potassium channel Kir5.1; Kcnj16; KIR5.1; MGC33717; potassium channel, inwardly rectifying subfamily J member 16; potassium channel, inwardly rectifying subfamily J, member 16; potassium inwardly rectifying channel subfamily J member 16; potassium inwardly-rectifying channel J16; potassium inwardly-rectifying channel, subfamily J, member 16; potassium voltage-gated channel subfamily J member 16
Gene Symbols Kcnj16
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human KCNJ16.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3773
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.