Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kir5.1 (KCNJ16) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575377
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575377 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575377 Supplier Thermo Scientific Supplier No. PA575377
Add to Cart
Add to Cart

Kir5.1 (KCNJ16) Polyclonal Antibody

Kir5.1 (KCNJ16) Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen Kir5.1 (KCNJ16)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Kcnj16
Gene Accession No. P52191, Q9NPI9, Q9Z307
Gene Alias 6430410F18Rik; AI132396; BIR9; Inward rectifier K(+) channel Kir5.1; inward rectifier K+ channel KIR5.1; inward rectifier potassium channel 16; inwardly-rectifying potassium channel Kir5.1; Kcnj16; KIR5.1; MGC33717; potassium channel, inwardly rectifying subfamily J member 16; potassium channel, inwardly rectifying subfamily J, member 16; potassium inwardly rectifying channel subfamily J member 16; potassium inwardly-rectifying channel J16; potassium inwardly-rectifying channel, subfamily J, member 16; potassium voltage-gated channel subfamily J member 16
Gene Symbols Kcnj16
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 376-418 of Human Kir5.1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16517, 29719, 3773
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.