Learn More
Kir6.1 Antibody (S366-60), DyLight 680, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259324FR
Catalog No. NBP259324FR
Only null left
Add to Cart
Description
Description
Kir6.1 Monoclonal specifically detects Kir6.1 in Human, Mouse, Rat, Gerbil samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Kir6.1
Monoclonal
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
KCNJ8
Fusion protein amino acids 306-424 (Cytoplasmic C-terminus) of rat Kir6.1
100 μL
3764
Human, Mouse, Rat, Gerbil
Purified
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
S366-60
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
ATP-sensitive inward rectifier potassium channel 8, Inward rectifier K(+) channel Kir6.1, inwardly rectifying potassium channel KIR6.1, KIR6.1, Potassium channel, inwardly rectifying subfamily J member 8, potassium inwardly-rectifying channel, subfamily J, member 8, uKATP-1
Mouse
Protein G purified
Primary
Detects 45kDa.
Store at 4C in the dark.
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.