Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kir6.1 (KCNJ8) Monoclonal Antibody (N366/60), PE
GREENER_CHOICE

Catalog No. PIMA545744
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545744 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545744 Supplier Invitrogen™ Supplier No. MA545744
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

A 1:100 dilution of MA5-45744 was sufficient for detection of Kir6.1 in 20 μg of mouse brain lysate by ECL immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 45kDa. This antibody was formerly sold as clone S366-60.

Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. KCNJ8 is an integral membrane protein and inward-rectifier type potassium channel. KCNJ8, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, is controlled by G-proteins.Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. The encoded protein, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, is controlled by G-proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kir6.1 (KCNJ8)
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N366/60
Concentration 1 mg/mL
Conjugate PE
Formulation 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.4
Gene KCNJ8
Gene Accession No. P97794, Q15842, Q63664
Gene Alias AI448900; ATP-sensitive inward rectifier potassium channel 8; ATP-sensitive inward rectifier potassium channel Kir6.1; gnite; Inward rectifier K(+) channel Kir6.1; inward rectifier potassium channel 6.1; inward rectifier potassium channel Kir6.1; inwardly rectifier K(+) channel Kir6.1; Inwardly rectifying potassium channel gene subfamily J-8 (ATP sensitive); Inwardly rectifying potassium channel gene, subfamily J-8 (ATP sensitive); inwardly rectifying potassium channel Kir6.1; ion channel; Kcnj8; KCNJ8 protein; Kir6.1; Kir6.1 like; potassium channel, inwardly rectifying subfamily J member 8; potassium channel, inwardly rectifying subfamily J, member 8; potassium inwardly rectifying channel subfamily J member 8; potassium inwardly-rectifying channel J8; potassium inwardly-rectifying channel, subfamily J, member 8; potassium voltage-gated channel subfamily J member 8; si:dkey-183c2.2; slmbr; sltr; UKATP1; uKATP-1; zkir6.1
Gene Symbols KCNJ8
Host Species Mouse
Immunogen Fusion protein amino acids 306-424 (Cytoplasmic C-terminus) of rat Kir6.1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16523, 25472, 3764
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.