Learn More
Kir6.2 (KCNJ11) Polyclonal Antibody, Invitrogen™
Guinea Pig Polyclonal Antibody
Supplier: Thermo Scientific PA5111821
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)SVAVAKA KPKFSIS Homology: Mouseamino acid residues identical; human - 12/14 amino acid residues identical.
This receptor is controlled by G proteins. Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium. Can be blocked by extracellular barium By similarity. Subunit of ATP-sensitive potassium channels (KATP). Can form cardiac and smooth muscle-type KATP channels with ABCC9. KCNJ11 forms the channel pore while ABCC9 is required for activation and regulation.
Specifications
Kir6.2 (KCNJ11) | |
Polyclonal | |
Unconjugated | |
Kcnj11 | |
AI842722; ATP-sensitive inward rectifier potassium channel 11; AW491124; BIR; HHF2; IKATP; Inward rectifier K(+) channel Kir6.2; IRK11; KATP; KATP channel; Kcnj11; Kir6.2; mBIR; MGC133230; MODY13; PHHI; Potassium channel, inwardly rectifying subfamily J member 11; TNDM3 | |
Guinea Pig | |
Antigen affinity chromatography | |
RUO | |
16514, 3767, 83535 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
P70673, Q14654, Q61743 | |
Kcnj11 | |
Peptide corresponding to amino acid residues 372-385 of rat Kir6.2 (intracellular, C-terminus) | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.