Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kir6.2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB094031 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094031 100 μL
NB094032 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094031 Supplier Novus Biologicals Supplier No. NBP303506100UL

Rabbit Polyclonal Antibody

Kir6.2 Polyclonal specifically detects Kir6.2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Kir6.2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias ATP-sensitive inward rectifier potassium channel 11, beta-cell inward rectifier subunit, BIRKIR6.2, HHF2, IKATP, Inward rectifier K(+) channel Kir6.2, inwardly rectifying potassium channel KIR6.2, Kir6.2, MGC133230, PHHI, potassium channel inwardly rectifing subfamily J member 11, Potassium channel, inwardly rectifying subfamily J member 11, potassium inwardly-rectifying channel, subfamily J, member 11, TNDM3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 171-390 of human Kir6.2 (NP_000516.3).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3767
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.