Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kir7.1 (KCNJ13) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103907
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103907 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103907 Supplier Thermo Scientific Supplier No. PA5103907
Add to Cart
Add to Cart

Kir7.1 (KCNJ13) Polyclonal Antibody

Kir7.1 (KCNJ13) Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen Kir7.1 (KCNJ13)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KCNJ13
Gene Accession No. O60928, O70617, P86046
Gene Alias Inward rectifier K(+) channel Kir7.1; inward rectifier potassium channel 13; Kcnj13; KIR1.4; KIR7.1; LCA16; Potassium channel, inwardly rectifying subfamily J member 13; potassium channel, inwardly rectifying subfamily J, member 13; potassium inwardly-rectifying channel, subfamily J, member 13; potassium voltage-gated channel subfamily J member 13; SVD
Gene Symbols KCNJ13
Host Species Rabbit
Immunogen A synthesized peptide derived from human KCNJ13(Accession O60928), corresponding to amino acid residues F60-F110.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100040591, 3769, 94341
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.