Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kir7.1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109875
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109875 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109875 Supplier Invitrogen™ Supplier No. PA5109875
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IARPKNRAFS IRFTDTAVVA HMDGKPNLIF QVANTRPSPL TSVRVSAVLY QERENGKLYQ TSVDFHLDGI SSDECPFFIF PLTYYHSITP SSPLATLLQH ENPSHFELVV FLSAMQEGTG EICQRRTSYL PSEIMLHHCF ASLLTRGSKG EYQIKMENFD KTVPEFPTPL VSKSPNRTDL DIHINGQSID NFQISETGLT E.

Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium. KCNJ13 has a very low single channel conductance, low sensitivity to block by external barium and cesium, and no dependence of its inward rectification properties on the internal blocking particle magnesium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kir7.1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene KCNJ13
Gene Accession No. O60928, P86046
Gene Alias Inward rectifier K(+) channel Kir7.1; inward rectifier potassium channel 13; Kcnj13; KIR1.4; KIR7.1; LCA16; Potassium channel, inwardly rectifying subfamily J member 13; potassium channel, inwardly rectifying subfamily J, member 13; potassium inwardly-rectifying channel, subfamily J, member 13; potassium voltage-gated channel subfamily J member 13; SVD
Gene Symbols KCNJ13
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 160-360 of human KCNJ13 (NP_002233.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100040591, 3769
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.