Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIRREL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543040
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543040 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543040 Supplier Invitrogen™ Supplier No. PA543040
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: FLEVGTLERY TVERTNSGSG VLSTLTINNV MEADFQTHYN CTAWNSFGPG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

KIRREL (NEPH1) is a member of the nephrin-like protein family, which includes NEPH2 and NEPH3. The cytoplasmic domains of these proteins interact with the C terminus of podocin (NPHS2), and the genes are expressed in kidney podocytes, cells involved in ensuring size- and charge-selective ultrafiltration. NEPH1 is a member of the nephrin-like protein family, which includes NEPH2. The cytoplasmic domains of these proteins interact with the C terminus of podocin (NPHS2), and the genes are expressed in kidney podocytes, cells involved in ensuring size- and charge-selective ultrafiltration (Sellin et al., 2003).
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIRREL
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KIRREL1
Gene Accession No. Q96J84
Gene Alias 6720469N11Rik; FLJ10845; kin of IRRE like (Drosophila); kin of IRRE like 1; Kin of irregular chiasm-like protein 1; kin of IRRE-like protein 1; Kirrel; KIRREL1; MGC129542; MGC129543; NEPH1; nephrin 1; nephrin related; nephrin-like protein 1
Gene Symbols KIRREL1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human KIRREL.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55243
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.