Learn More
Invitrogen™ KIRREL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589723
Description
Immunogen sequence: QGTQTRFSQE PADQTVVAGQ RAVLPCVLLN YSGIVQWTKD GLALGMGQGL KAWPRYRVVG SADAGQYNLE ITDAELSDDA SYECQATEAA LRSRRAKLTV LIPPEDTRID GGPVILLQAG TPHNLTCRAF NAKPAATIIW FRDGTQQEGA VASTELLKDG KRETTVSQLL INPTDLDIGR VFTC; Positive Samples: Mouse kidney, Mouse lung; Cellular Location: Cell membrane, Single-pass type I membrane protein.
KIRREL (NEPH1) is a member of the nephrin-like protein family, which includes NEPH2 and NEPH3. The cytoplasmic domains of these proteins interact with the C terminus of podocin (NPHS2), and the genes are expressed in kidney podocytes, cells involved in ensuring size- and charge-selective ultrafiltration. NEPH1 is a member of the nephrin-like protein family, which includes NEPH2. The cytoplasmic domains of these proteins interact with the C terminus of podocin (NPHS2), and the genes are expressed in kidney podocytes, cells involved in ensuring size- and charge-selective ultrafiltration (Sellin et al., 2003).
Specifications
KIRREL | |
Polyclonal | |
Unconjugated | |
Kirrel | |
6720469N11Rik; FLJ10845; kin of IRRE like (Drosophila); kin of IRRE like 1; Kin of irregular chiasm-like protein 1; kin of IRRE-like protein 1; Kirrel; KIRREL1; MGC129542; MGC129543; NEPH1; nephrin 1; nephrin related; nephrin-like protein 1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
170643, 310695 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.85 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q6X936, Q80W68 | |
Kirrel, KIRREL1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 17-200 of human KIRREL (NP_0607103). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.