Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kirrel1/NEPH1 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298231V
Description
Kirrel1/NEPH1 Polyclonal specifically detects Kirrel1/NEPH1 in Mouse samples. It is validated for ELISA.Specifications
Kirrel1/NEPH1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
55243 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
DyLight 405 | |
FLJ10845, kin of IRRE like (Drosophila), MGC129542, MGC129543, NEPH1, nephrin-like protein 1 | |
Produced in rabbits immunized with purified, recombinant Kirrel1/NEPH1 (NP_570937.2; Met1-Leu525). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction