Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIRREL2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987449
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7449 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7449 Supplier Bioss Supplier No. BS6721R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a type I transmembrane protein and member of the immunoglobulin superfamily of cell adhesion molecules. The encoded protein localizes to adherens junctions in pancreatic beta cells and regulates insulin secretion. Autoantibodies against the encoded protein have been detected in serum from patients with type 1 diabetes. This gene may also play a role in glomerular development and decreased expression of this gene has been observed in human glomerular diseases. This gene and the related opposite-strand gene nephrin are regulated by a bidirectional promoter.

Specifications

Antigen KIRREL2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KIRREL2
Gene Accession No. Q6UWL6
Gene Alias C330019F22Rik; DKFZp564A1164; FILTRIN; kin of IRRE like 2; kin of IRRE like 2 (Drosophila); kin of irregular chiasm-like 2; Kin of irregular chiasm-like protein 2; kin of IRRE-like 2; Kin of IRRE-like protein 2; kirre like nephrin family adhesion molecule 2; Kirrel2; MGC15718; Neph3; nephrin-like 3; nephrin-like gene 1; Nephrin-like protein 3; NLG1; UNQ5827/PRO19646; X kin of IRRE like 2
Gene Symbols KIRREL2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NLG1/KIRREL2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84063
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.