Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KISS1 Mouse anti-Human,Mouse, Unconjugated, Clone: 6A4.27, MilliporeSigma™

Catalog No. MABC17512MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC17512MI 25 μL
MABC17511MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC17512MI Supplier MilliporeSigma Supplier No. MABC175125UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KISS1 Monoclonal antibody specifically detects KISS1 in Human,Mouse samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KISS1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 6A4.27
Conjugate Unconjugated
Gene Accession No. NP_002247
Gene Alias Metastasis-suppressor KiSS-1;Kisspeptin-1
Gene Symbols KISS1, PP5098
Host Species Mouse
Immunogen A linear peptide corresponding to 54 amino acids from the C-terminal half of human Kisspeptin-1 (KISS1).
Molecular Weight of Antigen 14.70 kDa calculated.
Purification Method Protein G Purified
Quantity 25 μL
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Test Specificity Clone 6A4.27 is a mouse monoclonal antibody that detects Kisspeptin-1. It targets an epitope with in the KP54 (G68-F121) region.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.