Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KISS1R Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533879
Description
Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey (95%) Marmoset (80%).
GCPR54 is a galanin-like G protein coupled receptor that binds metastin, a peptide encoded by the metastasis suppressor gene KISS1. The tissue distribution of the protein suggests that it is involved in the regulation of endocrine function, and this is supported by the finding that it appears to play a role in the onset of puberty. GPR54 has been reported in various human brain regions, placenta, pancreas, and spinal cord, and at lower levels in spleen, peripheral blood leukocytes, testis, lymph node, thymus, small intestine, stomach, lung, and kidney. The receptor is also abundant in tumor tissues. In rat, GPR54 is expressed in liver, intestine, and in the pons, midbrain, thalamus, hypothalamus, hippocampus, amygdala, cortex (including frontal), and striatum. ESTs have been isolated from normal placenta and kidney cancer libraries.
Specifications
| KISS1R | |
| Polyclonal | |
| Unconjugated | |
| KISS1R | |
| AXOR12; CPPB1; G protein-coupled receptor 54; Gpr54; G-protein coupled receptor 54; G-protein coupled receptor OT7T175; HH8; HOT7T175; Hypogonadotropin-1; kiSS-; KiSS-1; KISS1 receptor; kiSS-1 receptor; KISS1R; KiSS-1R; Kisspeptins receptor; metastin receptor; mOT7T175; orphan G protein-coupled receptor 54; orphan G protein-coupled receptor GPR54; rOT7T175 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84634 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q969F8 | |
| KISS1R | |
| Synthetic 20 amino acid peptide from C-terminus of human KISS1R / GPR54. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction