Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kisspeptin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980484
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0484 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0484 Supplier Bioss Supplier No. BS0749R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene is a metastasis suppressor gene that suppresses metastases of melanomas and breast carcinomas without affecting tumorigenicity. The encoded protein may inhibit chemotaxis and invasion and thereby attenuate metastasis in malignant melanomas. Studies suggest a putative role in the regulation of events downstream of cell-matrix adhesion, perhaps involving cytoskeletal reorganization. A protein product of this gene, kisspeptin, stimulates gonadotropin-releasing hormone (GnRH)-induced gonadotropin secretion and regulates the pubertal activation of GnRH neurons. A polymorphism in the terminal exon of this mRNA results in two protein isoforms. An adenosine present at the polymorphic site represents the third position in a stop codon. When the adenosine is absent, a downstream stop codon is utilized and the encoded protein extends for an additional seven amino acid residues.

Specifications

Antigen Kisspeptin
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KISS1
Gene Accession No. Q15726
Gene Alias 0610012C01Rik; Eseptin; HH13; Kiss1; KiSS-1; KiSS-1 metastasis-suppressor; Kiss1 metastasis-suppressor; Kiss1 variant E1a-E2b-E3; Kiss1 variant E1b-E2a-E3; Kiss1 variant E1b-E2b-E3; kisspeptin; Kisspeptin-1; Kisspeptin-10; Kisspeptin-13; Kisspeptin-14; Kisspeptin-52; Kisspeptin-54; malignant melanoma metastasis-suppressor; Metastasis-suppressor KiSS-1; metastatin; Metastin; Metastin45-54; MLL septin-like fusion; PP5098; prepro-kisspeptin
Gene Symbols KISS1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Kiss-1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3814
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.