Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Easy-DNA™ gDNA Purification Kit

Catalog No. k180001
Encompass_Preferred
Click to view available options
Quantity:
1 Kit

Catalog No. K180001

Supplier: Invitrogen™ K180001

Only null left
Add to Cart
Add to Cart

Includes

The Easy-DNA Kit contains enough reagents for 15–200 samples, depending on sample size and type.This kit contains: 55mL Solution A (Lysis Solution), 25mL Solution B (Precipitation Solution), 100mL TE Buffer, 750μl Mussel Glycogen (2mg/mL in sterile water), 750 μl RNase (2mg/mL in sterile water), 750μl Protein Degrader (5mg/mL in sterile water)

The Easy-DNA™ Kit is fast and easy way to isolate high-quality, high-molecular weight genomic DNA (gDNA) from a wide range of cells and tissue types.

The Easy-DNA™ Kit is fast and easy way to isolate high-quality, high-molecular weight genomic DNA (gDNA) from a wide range of cells and tissue types. Sample sizes can range from single hair follicle up to 1 gram of mammalian tissue. Other samples tested include mouse tails, plant leaves, yeast, and E. coli cells. These samples yield high-quality DNA with average size between 100–200 kb, which is suitable for PCR, DNA hybridization, genomic DNA library construction, and other applications.

  • Reliability: High-quality genomic DNA from large or small samples of cells, tissue, plant, yeast, E. coli, blood, and even hair follicles
  • Simplicity: For most sample types, only 4 steps, typically requiring less than 90 minutes are needed to isolate high-quality DNA
  • Versatility: One kit is supported by optimized protocols for variety of cell and tissue types

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 55 mL Solution A; room temperature
• 25 mL Solution B; room temperature
• 100 mL TE Buffer; room temperature
• 750 μL Mussel Glycogen; room temperature, or -20°C for long-term storage
• 750 μL RNase; room temperature, or -20°C for long-term storage
• 750 μL Protein Degrader; room temperature, or -20°C for long-term storage

Isolation Technology Organic Extraction
Sample Type Bacteria, Cells, Hair Follicles, Mouse Tails, Plant, Tissue, Virus, Whole Blood, Yeast
Elution Volume 10 to 150 μL
Final Product Type gDNA
For Use With (Application) PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization
High-throughput Compatibility Not High-throughput Compatible (Manual)
Quantity 1 Kit
Shipping Condition Room Temperature
Test Time Up to 90 min.
Yield ≤850 μg
Starting Material Amount Bacteria: ≤109 cells
Cells: ≤108
Hair Follicles: ≤5
Mouse Tails: 1 cm
Plant: ≤50 mg
Tissue: ≤1 g
Virus: ≤750 μL
Whole Blood: ≤2 mL
Yeast: ≤10 mL
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.