Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIT Mouse anti-Human, Unconjugated, Clone: KIT/983, Abnova™

Catalog No. 89337759 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-759 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-759 Supplier Abnova Corporation Supplier No. MAB21268
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

KIT Monoclonal antibody specifically detects KIT in Human samples. It is validated for IHC-P, IF, Flow Cyt.

Specifications

Antigen KIT
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone KIT/983
Conjugate Unconjugated
Description v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog
Gene KIT
Gene Alias C-Kit, CD117, PBT, SCFR
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 100-300 of human KIT
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3815
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.