Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kjeldahl Digestion Reagent with Copper Sulfate, APHA 4500-N(org) B, Reagents

Catalog No. 01334855
Click to view available options
Quantity:
1 L
4 L

Catalog No. 01-334-855

Supplier: Reagents Holdings Llc CK2004004A

Only null left
Add to Cart
Add to Cart

Kjeldahl Digestion Reagent with Copper Sulfate, APHA 4500-N(org) B, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Boiling Point 102°C
Color Blue
CAS 7778-80-5, 7664-93-9, 7732-18-5
Physical Form Liquid
Quantity 4 L
Packaging Poly Bottle
Grade Reagent
Chemical Name or Material Kjeldahl Digestion Reagent with Copper Sulfate, APHA 4500-N(org) B
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.