Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kjeldahl Solution, 50% Sodium Hydroxide/2.5% Sodium Thiosulfate, Reagents

Catalog No. 01337211
Change view
Click to view available options
Quantity:
4 L
20 L
Catalog No. Quantity
01-337-211 20 L
01-337-210 4 L
2 options

Catalog No. 01-337-211

Supplier: Reagents Holdings Llc C222005020F

Only null left
Add to Cart
Add to Cart

Kjeldahl Solution, 50% Sodium Hydroxide/2.5% Sodium Thiosulfate, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Kjeldahl Solution, 50% Sodium Hydroxide/2.5% Sodium Thiosulfate
Melting Point -7°C
Boiling Point 102°C
Color Colorless
CAS 10102-17-7, 1310-73-2, 7732-18-5
Physical Form Liquid
Quantity 20 L
Packaging Cubitainer
Synonym Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.