Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI Kjeldahl Tablets
SDP

Catalog No. p-7219101
Click to view available options
Chemical Name or Material:
Antifoam
Missouri
Titanium

Digestion aids for nitrogen determination.

  • Environmentally friendly: free of toxic mercury and selenium
  • Different tablets to fit individual needs
  • For digestion

Specifications

Chemical Name or Material Antifoam
Melting Point 1623°C (Main Component)
Color White
Quantity 1000 Pk.
CAS 7757-82-6, 7631-86-9
Composition 0.97 g Na2SO4, 0.03 g Silicone Antifoam
Packaging Plastic Container
Synonym None
Percent Purity Mixture
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.