Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kjeltabs S-3.5, Foss™
Click to view available options
Quantity:
1000 Tablets
Specifications
Specifications
Physical Form | Flat Round Speckled Tablets |
CAS | 7782-49-2 |
Quantity | 1000 Tablets |
Chemical Name or Material | Selenium Kjeltabs |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction