Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLC1 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP146840C
Description
KLC1 Polyclonal antibody specifically detects KLC1 in Human samples. It is validated for Western BlotSpecifications
KLC1 | |
Polyclonal | |
Western Blot | |
hKLC1B, hKLC1G, hKLC1J, hKLC1S, kinesin 2, kinesin 2 60/70kDa, kinesin light chain 1, KLC 1, KLChKLC1N, KNS2AhKLC1P, KNS2hKLC1R, medulloblastoma antigen MU-MB-2.50, MGC15245 | |
The immunogen for this product maps to a region between residue 523 and 573 of human kinesin light chain 1 using the numbering given in entry NP_891553.2 (GeneID 3831). | |
0.1 mL | |
Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
3831 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction