Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596356
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596356 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596356 Supplier Invitrogen™ Supplier No. PA596356
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LALVYRDQNK YKDAANLLND ALAIREKTLG KDHPAVAATL NNLAVLYGKR GKYKEAEPLC KRALEIREKV LGKDHPDVAK QLNNLALLCQ NQGKYEEVEY YYQRALEIYQ TKLGPDDPNV AKTKNNLASC YLKQGKFKQA ETLYKEILTR AHEREFGSVD DENKPIWMHA EEREECKGKQ KDGTSFGEYG GWYKACKVDS PTVTTTLKNL GALYRRQGKF EAAETLEEAA MRSRKQGLDN VHKQRVAEVL NDPENMEKRR SRESLNVDVV KYESGPDGGE EVSMSVEWNG MRKMKLGLVN; Positive Samples: SW480, 22Rv1, BT-474, MCF7, Mouse brain; Cellular Location: Cell projection, Cytoplasm, Cytoplasmic vesicle, cytoskeleton, growth cone.

Conventional kinesin is a tetrameric molecule composed of two heavy chains and two light chains, and transports various cargos along microtubules toward their plus ends. The heavy chains provide the motor activity, while the light chains bind to various cargos. This gene encodes a member of the kinesin light chain family. It associates with kinesin heavy chain through an N-terminal domain, and six tetratricopeptide repeat (TPR) motifs are thought to be involved in binding of cargos such as vesicles, mitochondria, and the Golgi complex. Thus, kinesin light chains function as adapter molecules and not motors per se. Although previously named kinesin 2, this gene is not a member of the kinesin-2 / kinesin heavy chain subfamily of kinesin motor proteins. Extensive alternative splicing produces isoforms with different C-termini that are proposed to bind to different cargos; however, the full-length nature of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLC1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KLC1
Gene Accession No. O88447, P37285, Q07866
Gene Alias AI874768; kinesin 2; kinesin 2 60/70kDa; kinesin light chain 1; Kinesin2; Klc; KLC 1; Klc1; KNS 2; KNS 2A; Kns2; KNS2A; medulloblastoma antigen MU-MB-2.50; MGC15245
Gene Symbols KLC1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 261-560 of human Kinesin light chain 1 (Kinesin light chain 1 (KLC1)) (NP_0055432).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16593, 171041, 3831
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.