Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLC2 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300308UV
Description
KLC2 Polyclonal specifically detects KLC2 in Human samples. It is validated for ELISA.Specifications
KLC2 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
64837 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
DyLight 350 | |
FLJ12387, kinesin light chain 2, KLC 2 | |
Produced in rabbits immunized with purified, recombinant KLC2 (NP_073733.1; Met1-Gly622). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction