Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114004
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114004 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114004 Supplier Invitrogen™ Supplier No. PA5114004
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLCQNQGKFE DVERHYARAL SIYEALGGPH DPNVAKTKNN LASAYLKQNK For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 86%; Guinea Pig: 86%; Horse: 92%; Human: 100%; Mouse: 100%; Rabbit: 77%; Rat: 100%.

KLC3 encodes a member of the kinesin light chain gene family. Kinesins are molecular motors involved in the transport of cargo along microtubules, and are composed of two kinesin heavy chain (KHC) and two kinesin light chain (KLC) molecules. KLCs are thought to typically be involved in binding cargo and regulating kinesin activity. In the rat, a protein similar to this gene product is expressed in post-meiotic spermatids, where it associates with structural components of sperm tails and mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLC3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KLC3
Gene Accession No. Q6P597
Gene Alias BC017147; kinesin light chain 2; kinesin light chain 3; Kinesin light chain KLCt; KLC2; KLC2L; KLC2-like; Klc3; Klct; KNS2B
Gene Symbols KLC3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human KLC3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147700
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.