Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLDC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556359
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556359 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556359 Supplier Invitrogen™ Supplier No. PA556359
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DTVLLWGGRN DTEGACNVLY AFDVNTHKWF TPRVSGTVPG ARDGHSACVL GKIMYIFGGY EQQADCFSND IHKLDTSTMT WTLICTKGSP ARWRD Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.

The BTB (Broad-Complex, Tramtrack and Bric a brac) domain, also known as the POZ (Poxvirus and Zinc finger) domain, is an N-terminal homodimerization domain that contains multiple copies of kelch repeats and/or C2H2-type zinc fingers. Proteins that contain BTB domains are thought to be involved in transcriptional regulation via control of chromatin structure and function. The Kelch domain-containing protein 3 (KLHDC3), also designated PEAS, contains 5 Kelch repeats and may be involved in meiotic recombination process. The gene encoding KLHDC3 maps to chromosome 6, which contains around 1, 200 genes within 170 million base pairs of sequence. Deletion of a portion of the q arm of chromosome 6 is associated with early onset intestinal cancer suggesting the presence of a cancer susceptibility locus. Porphyria cutanea tarda, Parkinson's disease, Stickler syndrome, 21-hydroxylase deficiency and maple syrup urine disease are also associated with genes on chromosome 6. KLHDC3 protein is mainly expressed in cytoplasm, also found in meiotic chromatin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLDC3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KLHDC3
Gene Accession No. Q9BQ90
Gene Alias 1300011D16Rik; dJ20C7.3; kelch domain containing 3; kelch domain-containing protein 3; KLHDC3; Peas; Protein Peas; RP1-20C7.3; testis intracellular mediator protein
Gene Symbols KLHDC3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KLDC3. Recombinant protein control fragment (Product #RP-94800).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116138
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.