Learn More
Invitrogen™ KLDC3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589401
Description
Immunogen sequence: MLRWTVHLEG GPRRVNHAAV AVGHRVYSFG GYCSGEDYET LRQIDVHIFN AVSLRWTKLP PVKSAIRGQA PVVPYMRYGH STVLIDDTVL LWGGRNDTEG ACNVLYAFDV NTHKWFTPRV SGTVPGARDG HSACVLGKIM YIFGGYEQQA DCFSNDIHKL DTSTMTWTLI CTKGSPARWR DFHSATMLGS HMYVFGGRAD RFGPFHSNNE IYCNRIRVFD TRTEAWLDCP PTPVLPEGRR SHSAFGYNGE LYIFGGYNAR LNRHFHDLWK FNPVSFTWKK IEPKGKGPCP RRRQCCCIVG DKIVLFGGTS PSPEEGLGDE FDLIDHSDLH ILDFSPSLKT LCKLAVIQYN LDQSCLPHDI RWELNAMTTN SNISRPIVSS HG; Positive Samples: H460, DU145, HepG2; Cellular Location: Cytoplasm.
The BTB (Broad-Complex, Tramtrack and Bric a brac) domain, also known as the POZ (Poxvirus and Zinc finger) domain, is an N-terminal homodimerization domain that contains multiple copies of kelch repeats and/or C2H2-type zinc fingers. Proteins that contain BTB domains are thought to be involved in transcriptional regulation via control of chromatin structure and function. The Kelch domain-containing protein 3 (KLHDC3), also designated PEAS, contains 5 Kelch repeats and may be involved in meiotic recombination process. The gene encoding KLHDC3 maps to chromosome 6, which contains around 1, 200 genes within 170 million base pairs of sequence. Deletion of a portion of the q arm of chromosome 6 is associated with early onset intestinal cancer suggesting the presence of a cancer susceptibility locus. Porphyria cutanea tarda, Parkinson's disease, Stickler syndrome, 21-hydroxylase deficiency and maple syrup urine disease are also associated with genes on chromosome 6. KLHDC3 protein is mainly expressed in cytoplasm, also found in meiotic chromatin.
Specifications
KLDC3 | |
Polyclonal | |
Unconjugated | |
KLHDC3 | |
1300011D16Rik; dJ20C7.3; kelch domain containing 3; kelch domain-containing protein 3; KLHDC3; Peas; Protein Peas; RP1-20C7.3; testis intracellular mediator protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
116138 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.17 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q9BQ90 | |
KLHDC3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-382 of human KLHDC3 (NP_4765021). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.