Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Klebsiella aerogenes K15 Monoclonal Antibody (ZX3.1)
GREENER_CHOICE

Catalog No. PIMA518038
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518038 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518038 Supplier Invitrogen™ Supplier No. MA518038
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody reacts with Klebsiella aerogenes serotype 15.

Klebsiella are non-motile, rod-shaped, aerobic bacteria that possess a prominent polysaccharide capsule. Some of these bacteria produce an extracellular toxic complex that has been shown to be 'lethal for and produce extensive lung pathology in mice'. It is composed of 63% capsular polysaccharide, 30% lipopolysaccharide, and 7% protein; when introduced to experimental animals in sublethal doses, the animals built up immunization due to antibody production. Klebsiella pneumoniae is one of the most common Gram-negative bacteria seen by physicians worldwide. Pneumonias that are caused by Klebsiella pneumoniae are difficult to control; mortality rates have even been reported as up to 50% after antibiotic treatment. Nitrogen metabolism in Klebsiella aerogenes has also been studied throroughly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Klebsiella aerogenes K15
Applications ELISA
Classification Monoclonal
Clone ZX3.1
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 10 mM sodium azide
Gene Alias k aerogenes; K. aerogenes
Host Species Mouse
Immunogen LPS membrane extract from K.aerogenes.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.