Learn More
Invitrogen™ Klebsiella aerogenes K21 Monoclonal Antibody (1231/153)

Description
Klebsiella aerogenes K21 Monoclonal Antibody for ELISA
Specifications
Specifications
| Antigen | Klebsiella aerogenes K21 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 1231/153 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene Alias | Enterobacter aerogenes |
| Host Species | Mouse |
| Immunogen | K. aerogenes purified polysaccharide K21. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.