Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Klebsiella aerogenes Monoclonal Antibody (57/82)
GREENER_CHOICE

Catalog No. PIMA546607
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546607 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA546607 Supplier Invitrogen™ Supplier No. MA546607
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Klebsiella aerogenes Monoclonal Antibody for ELISA

Klebsiella aerogenes, previously known as Enterobacter aerogenes, is a Gram-negative, oxidase negative, catalase positive, citrate positive, indole negative, rod-shaped bacterium. It is composed of 63% capsular polysaccharide, 30% lipopolysaccharide, and 7% protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Klebsiella aerogenes
Applications ELISA
Classification Monoclonal
Clone 57/82
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Alias Enterobacter aerogenes
Host Species Mouse
Immunogen A crude sonicate of Klebsiella aerogenes.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 λ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.