Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ A40 Liquid and Particle Protection Boot Covers
GREENER_CHOICE

Catalog No. 19057521 Shop All Ansell Products
Change view
Click to view available options
Size:
Medium/Large
Universal
X-Large/2X-Large
Height (English):
17 in.
18 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Height (English)
19-057-521 Medium/Large 18 in.
19-120-2412 Universal 17 in.
19-121-718 X-Large/2X-Large 18 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-057-521 Supplier Ansell Supplier No. 36779
Add to Cart
Add to Cart
This item is not returnable. View return policy

A40 boot covers feature a highly durable PVC sole and are made with breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.

  • Boot Covers available in 3 sizes, choose from 17 in. or 18 in. height
  • Elastic opening at top and elastic ankle
  • Serged seams
  • Highly durable PVC sole
  • Made to keep out debris, dry particulates, and liquid splashes
  • Adheres to NFPA 99 criteria for antistatic materials

Compliance

Meets NFPA™ 99 Health Care Facilities criteria for antistatic materials.

TRUSTED_SUSTAINABILITY

Specifications

Size Medium/Large
Material Microporous Fabric
Coating Microporous Film Laminate
Color White
Sterility Non-sterile
Closure Type Elastic Top and Ankles
Sole Type PVC
Model 36779
Seam Type Serged
For Use With (Application) Aviation, Pharmaceutical Manufacturing, Liquid Handling, General Manufacturing
Certifications/Compliance NFPA 99, ASTM 1670
Packaging Quantity 400/Cs.
Height (English) 18 in.
Height (Metric) 45.7 cm
Resistance Fluid
Product Type Boot Covers
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.