Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ KGA300 Coveralls with Hood
GREENER_CHOICE

Catalog No. 17004488 Shop All Ansell Products
Change view
Click to view available options
Size:
2X-Large
3X-Large
4X-Large
5X-Large/6X-Large
Large
Medium
Small
X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-004-488 4X-Large
17-004-482 Small
17-004-483 Medium
17-004-484 Large
17-004-485 X-Large
17-004-486 2X-Large
17-004-487 3X-Large
17-004-489 5X-Large/6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 17-004-488 Supplier Ansell Supplier No. 41509
Add to Cart
Add to Cart
This item is not returnable. View return policy

KGA300 coveralls for surface preparation and paint protection are designed to protect against hazardous particulates and certain light liquid splashes while providing a comfortable fit and freedom of movement.

  • KGA300 liquid and surface prep and paint protection coveralls help protect against hazardous particulates and certain light liquid splashes
  • Made of disposable, lightweight microporous film laminate
  • Pass NFPA 99 for antistatic materials
  • Feature serged seams and a water resistant front zipper, and come in a variety of sizes and configurations
  • Human-centric Reflex™ design offers comfortable fit and freedom of movement
  • Eligible for recycling via RightCycle™ when used in non-hazardous applications as classified by federal, state, and local regulations
  • Zipper front, elastic wrists, ankles, and hood
  • Hood is designed to fit with respirator
  • Silicone- and latex-free
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size 4X-Large
Gender Unisex
Material Microporous Film Laminate
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Elastic
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Anti-static, Liquid, Particle
Certifications/Compliance NFPA 99
Latex Free Yes
Model KleenGuard™ KGA300
For Use With (Application) Surface Preparation and Painting
Quantity 25/Cs.
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.