Learn More
Ansell KleenGuard™ Calico Safety Glasses
Catalog No. 19170104 Shop All Ansell Products
Change view
Click to view available options
Lens Tint:
Amber
Smoke
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-170-104 Amber
17-000-253 Smoke
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-170-104 Supplier Ansell Supplier No. 25674
This item is not returnable. View return policy
KleenGuard™ Calico Safety Eyewear are a lightweight with wrap around and anti-fog protection in a stylish and comfortable design.
  • Wraparound design and impact-resistant lenses
  • Interchangeable head strap and temples allow customizatio for a closer fit or enhanced comfort
  • Lenses feature EN166 N anti-fog coating and fog shield
  • Cushioned brow bar for additional comfort and ventilation
  • Lens options include:
    • Amber anti-fog lenses with light gathering properties ideal for low-light and low-contrast environments
    • Smoke anti-fog lenses ideal for bright, sunny days to reduce glare
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Impact-resistant, UV-treated
Certifications/Compliance EN166 1FT, CSA, ANSI Z87.1+
Lens Tint Amber
For Use With (Application) Areas where there's a high risk for airborne debris
Frame Color Black
No. per Case 12 pairs/Cs.
Lens Material Polycarbonate
Frame Material Plastic with foam brow bar
UV Protection 0.999
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.