Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KleenGuard™ G80 Neoprene/Latex Chemical Resistant Gloves

Catalog No. 19006228 Shop All Ansell Products
Change view
Click to view available options
Size:
Large
Medium
Small
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-006-228 Small
19-006-231 X-Large
19-006-229 Medium
19-006-230 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-006-228 Supplier KleenGuard™ Supplier No. 38741
Add to Cart
Add to Cart
This item is not returnable. View return policy

G80 gloves have a natural rubber latex construction covered in neoprene to give a high resistance to acids, alkalis, oils and solvents. Ideal for removing paint, sealants and residues from a surface or degreasing machinery with a strong solvent.

  • Neoprene over natural rubber latex construction with cotton flock lining for comfort and perspiration absorption
  • High resistance to acids, oils, solvents and alkalis
  • Meets the EN 374-AKL standard and has a high level of dexterity, per EN420:2003 standard
  • Not suitable for those with Type 1 latex allergies
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003, EN 374
Applications Industrial
Color Blue, Yellow
Latex Free No
Material Neoprene, Latex
Product Type Chemical-resistant Gloves
Size Small
Thickness (English) 27.5 mil
Thickness (Metric) 0.69 mm
EN ISO 374-1 CHEMICAL LETTER CODES AKL
Glove Cuff Style Straight
Grip Design Textured Fingertips
Liner Material Cotton Flock
Lining Lined
Model 38741
Series Kleenguard™ G80
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.