Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ G10 Kraken Grip™ Black Nitrile Glove
GREENER_CHOICE

Catalog No. p-7161442
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Black nitrile gloves feature tread-like texture that channels oil, grease and solvents away from the areas of the hand where grip is most critical. Provides powder free moisture control on the inside for user comfort.

  • Fully textured to give technicians dexterity and control when performing delicate tasks
  • Powder free
  • Ambidextrous
  • Approved for food handling, per US FDA regulations Health Canada food regulations
  • Beaded cuffs prevent roll-down to help alleviate contamination concerns
  • Latex-free
  • California Prop 65 compliant
  • Typical Applications:
    • Fabrication/machining
    • Part handling
    • Assembly
    • Indoors/outdoors maintenance
    • Cleaning
    • Chemical splash
    • Food handling
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CFIA Approved, California Prop 65 Compliant
Chemotherapeutic Rated No
Applications Industrial
AQL 1.5
Color Black
Disposable Single-use
Exterior Finish Fully Textured
Finger Thickness (English) 6 mil
Finger Thickness (Metric) 0.15 mm
Laboratory/Cleanroom Use No
Latex Free Yes
Material Nitrile
Palm Thickness (English) 4.7 mil
Palm Thickness (Metric) 0.12 mm
Powdered Powder Free
Size Small
Thickness (English) 6 mil
Thickness (Metric) 0.15 mm
Model 49275
Series Kleenguard™ G10 Kraken Grip™
Length (English) 9.5 in.
Length (Metric) 24.13 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.