Learn More
Ansell KleenGuard™ Nemesis™ CSA Safety Glasses
KleenGuard™ Nemesis™ CSA Safety Glasses feature durable lightweight construction for protection at work and at home.
Supplier: Ansell 20385
View more versions of this product
Description
- CSA certified and meets ANSI Z87.1+ standards for personal eye protection
- Sporty, flexible, lightweight design
- Polycarbonate lenses provide 99.9% UVA/ UVB/UVC protection
- FLEX-DRY™ nosepiece is designed to be extremely flexible, to channel sweat away from the eyes, and to help prevent slippage down the nose
- Include several lens options:
- Bronze lenses provide visual definition with increased contrast and enhanced contours in most light conditions
- Light blue lenses provide enhanced contrast in sodium vapor lighting environments and bright work environments that use yellow incandescent/fluorescent lamps
- Copper blue shield lenses have a light brown tint and are designed to filter out blue light for enhanced sharpness and depth perception; ideal for low-light conditions
- Smoke lenses are worn on bright, sunny days to provide glare reduction; ideal for outdoor use
- Amber/yellow lenses block out blue light, making the environment appear brighter; ideal for low-light conditions and indoor use
Specifications
Impact-resistant, UV-treated | |
Smoke | |
Gun Metal | |
Polycarbonate | |
UV400 |
ANSI Z87.1+, CSA Z94.3 | |
Safety sunglasses | |
12 pairs/Cs. | |
Plastic |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.