Description
- Includes neck cord to keep glasses safe when not being worn
- Extra-lightweight frameless design
- No-brow design increases upward peripheral vision and limits interference with hard hat suspension systems
- Meets ANSI Z87.1+ standards for personal eye protection
- TAA compliant
- 8.25 base curve polycarbonate lenses provide 99.9% UVA/ UVB/UVC protection
- Lightweight and frameless for long-term comfort
- Recyclable via The RightCycle Program
- Several lens options including:
- Bronze lenses provide visual definition with increased contrast
- IRUV Shade 5 lenses protect against infrared (IR) and ultraviolet radiation (UVA/UVB)
Specifications
Specifications
| Lens Coating | Impact-resistant, UV-treated |
| Certifications/Compliance | ANSI Z87.1+ |
| Lens Tint | Bronze, Indoor/Outdoor |
| For Use With (Application) | General-purpose eye protection |
| Frame Color | Camouflage |
| No. per Case | 12 pairs/Cs. |
| Lens Material | Polycarbonate |
| Frame Material | Plastic |
| UV Protection | 0.999 |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.