Learn More
Ansell KleenGuard™ V40 Maxfire™ Small Safety Eyewear
Same contemporary styling as the KleenGuard Maxfire safety glasses, but smaller to fit smaller, slimmer heads. Flexible click-to-fit temples and adjustable nose pad allows for fit and lens angle to be customized for user comfort.
Specifications
Lens Coating Anti-fog
Product Type Safety Eyewear
View More Specs
Products 4
Catalog Number Mfr. No. Frame Color Frame Type Lens Coating Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. Frame Color Frame Type Lens Coating Lens Tint Price Quantity & Availability  
19-006-201
View Documents
Ansell
38484
Brown Brown frame with brown tips Anti-fog Smoke N/A N/A N/A
19-006-203
View Documents
Ansell
38493
Black Black frame with red tips Anti-fog Amber N/A N/A N/A
19-006-202
View Documents
Ansell
38489
Brown Brown frame with brown tips Anti-fog Indoor/Outdoor N/A N/A N/A
19-006-200
View Documents
Ansell
38498
Black Black frame with red tips Anti-fog Clear N/A N/A N/A
Description
Description
  • Wearers can adjust the angle of the lens for each work application
  • Glsses feature 5-position click-to-fit temples for a personalized fit
  • Built-in comfort features including an adjustable nose pad and soft rubber inserts on the brow, temples and along the ear
  • Premium anti-scratch (EN 166 K Rating) and anti-fog (EN 166 N Rating) properties
  • Metal-free design (with screw-less hinges) helps minimize electrical transference risk
  • Meet ANSI Z87.1+ standards for high impact
  • Polycarbonate lenses provide 99.9% UVA / UVB / UVC protection
Specifications
Specifications
Anti-fog
Safety Eyewear
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.