Description
- Three-point extendable temples help give the wearer a customized fit and enhanced comfort
- ANSI Z87.1+ standard for impact protection and the polycarbonate lenses provide 99.9% UVA / UVB / UVC protection
- Hard coated lenses for longer life and scratch protection
- Recyclable via The RightCycle Program
Specifications
Specifications
| Lens Coating | Anti-fog, Impact-resistant, UV-treated |
| Certifications/Compliance | ANSI Z87.1+ |
| Lens Tint | Smoke |
| For Use With (Application) | General-purpose eye protection |
| Frame Color | Black, Gray |
| No. per Case | 12 pairs/Cs. |
| Lens Material | Polycarbonate |
| Frame Material | Plastic |
| UV Protection | UV400 |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.