Learn More
Ansell KleenGuard™ Visio™ Safety Glasses
GREENER_CHOICE
Catalog No. 19005062 Shop All Ansell Products
Change view
Click to view available options
Frame Color:
Black
Black, Gray
Clear
Frame Material:
Plastic
Polycarbonate
Lens Coating:
Anti-fog, Impact-resistant, UV-treated
Impact-resistant, Scratch-resistant, UV-treated
Lens Tint:
Amber
Clear
Mirror
Smoke
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Frame Material Lens Coating Lens Tint
19-005-062 Black Plastic Impact-resistant, Scratch-resistant, UV-treated Mirror
19-804-257 Clear Polycarbonate Anti-fog, Impact-resistant, UV-treated Clear
19-005-060 Black, Gray Plastic Anti-fog, Impact-resistant, UV-treated Smoke
19-005-061 Black Plastic Impact-resistant, Scratch-resistant, UV-treated Amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-005-062 Supplier Ansell Supplier No. 14475
Add to Cart
Add to Cart
This item is not returnable. View return policy
Visio™ Safety Glasses feature three-point extendable temples for a customized fit and enhanced comfort.
  • Three-point extendable temples help give the wearer a customized fit and enhanced comfort
  • ANSI Z87.1+ standard for impact protection and the polycarbonate lenses provide 99.9% UVA / UVB / UVC protection
  • Hard coated lenses for longer life and scratch protection
  • Recyclable via The RightCycle Program
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Impact-resistant, Scratch-resistant, UV-treated
Certifications/Compliance ANSI Z87.1+
Lens Tint Mirror
For Use With (Application) General-purpose eye protection
Frame Color Black
No. per Case 12 pairs/Cs.
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.