Learn More
KLF10 Polyclonal Antibody, Invitrogen™
Description
Recommended positive controls: HeLa nuclear, eGFP-human KLF10-transfected 293T. Predicted reactivity: Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | KLF10 |
| Applications | ChIP Assay, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | KLF10 |
| Gene Accession No. | Q13118 |
| Gene Alias | AI115143; Cpg20; early growth response-alpha; EGR[a]; EGRA; Egral; EGR-alpha; Gdnfif; GDNF-inducible factor; Klf10; Krueppel-like factor 10; Kruppel like factor 10; Kruppel-like factor 10; mGIF; TGFB inducible early growth response; TGFB inducible early growth response 1; TGFB-inducible early growth response protein 1; Tieg; TIEG1; TIEG-1; transcription factor GIF; Transforming growth factor-beta-inducible early growth response protein 1; zinc finger transcription factor homolog CPG20; zinc finger transcription factor TIEG |
| Gene Symbols | KLF10 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.