Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLF11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599200
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599200 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599200 Supplier Invitrogen™ Supplier No. PA599200
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KLF11.

The protein encoded by this gene is a zinc finger transcription factor that binds to SP1-like sequences in epsilon- and gamma-globin gene promoters. This binding inhibits cell growth and causes apoptosis. Defects in this gene are a cause of maturity-onset diabetes of the young type 7 (MODY7). Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF11
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KLF11
Gene Accession No. O14901, Q8K1S5
Gene Alias 9830142A17; D12Ertd427e; FKLF; FKLF1; klf11; Krueppel-like factor 11; Kruppel like factor 11; Kruppel-like factor 11; MODY7; Tcfcp2l2; TGFB inducible early growth response 2; TGFB inducible early growth response 3; TGF-beta inducible early growth response protein 2; TGFB-inducible early growth response protein 2; TGFB-inducible early growth response protein 2b; TGFB-inducible early growth response protein 3; Tieg2; TIEG-2; Tieg2b; Tieg3; TIEG-3; transcription factor CP2-like 2; transforming growth factor-beta-inducible early growth response protein 2; Transforming growth factor-beta-inducible early growth response protein 3
Gene Symbols KLF11
Host Species Rabbit
Immunogen A synthesized peptide derived from human KLF11(Accession O14901), corresponding to amino acid residues M1-T36.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 194655, 313994, 8462
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.