Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KLF11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116877
Description
Positive Samples: mouse thymus, mouse brain, rat lung Immunogen sequence: RAVDIMDICE SILERKRHDS ERSTCSILEQ TDMEAVEALV CMSSWGQRSQ KGDLLRIRPL TPVSDSGDVT TTVHMDAATP ELPKDFHSLS TLCIT.
The protein encoded by this gene is a zinc finger transcription factor that binds to SP1-like sequences in epsilon- and gamma-globin gene promoters. This binding inhibits cell growth and causes apoptosis. Defects in this gene are a cause of maturity-onset diabetes of the young type 7 (MODY7). Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
KLF11 | |
Polyclonal | |
Unconjugated | |
KLF11 | |
9830142A17; D12Ertd427e; FKLF; FKLF1; klf11; Krueppel-like factor 11; Kruppel like factor 11; Kruppel-like factor 11; MODY7; Tcfcp2l2; TGFB inducible early growth response 2; TGFB inducible early growth response 3; TGF-beta inducible early growth response protein 2; TGFB-inducible early growth response protein 2; TGFB-inducible early growth response protein 2b; TGFB-inducible early growth response protein 3; Tieg2; TIEG-2; Tieg2b; Tieg3; TIEG-3; transcription factor CP2-like 2; transforming growth factor-beta-inducible early growth response protein 2; Transforming growth factor-beta-inducible early growth response protein 3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
194655, 313994 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.97 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q8K1S5 | |
KLF11 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 13-107 of human KLF11 (NP_0035881). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction