Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF12 Antibody (1230C), HRP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB9097H
Description
KLF12 Monoclonal antibody specifically detects KLF12 in Human samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
KLF12 | |
Monoclonal | |
HRP | |
PBS | |
Rabbit | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
11278 | |
Store at 4C in the dark. |
Flow Cytometry, Immunocytochemistry | |
1230C | |
Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
AP-2rep, AP-2rep transcription factor, AP2REPAP-2 repressor, HSPC122, KLF12 zinc finger transcriptional repressor, Krueppel-like factor 12, Kruppel-like factor 12, Transcriptional repressor AP-2rep | |
Mouse myeloma cell line NS0-derived human KLF12, Asn2-Val402, Accession # Q9Y4X4 | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction