Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF12 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP32139125UL
Description
KLF12 Polyclonal antibody specifically detects KLF12 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
KLF12 | |
Polyclonal | |
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
AP-2rep, AP-2rep transcription factor, AP2REPAP-2 repressor, HSPC122, KLF12 zinc finger transcriptional repressor, Krueppel-like factor 12, Kruppel-like factor 12, Transcriptional repressor AP-2rep | |
This antibody was developed against Recombinant Protein corresponding to amino acids: SSPTVITSVSSASSSSTVLTPGPLVASASGVGGQQFLHIIHPVPPSSPMNLQSNKLSHVHRIPVVVQSVPVVYTAVRSPGNV | |
25 μg | |
DNA replication Transcription Translation and Splicing | |
11278 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction