Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF12 Antibody (PCRP-KLF12-1E3), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314277V
This item is not returnable.
View return policy
Description
KLF12 Monoclonal antibody specifically detects KLF12 in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
KLF12 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunofluorescence | |
PCRP-KLF12-1E3 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
AP-2rep, AP-2rep transcription factor, AP2REPAP-2 repressor, HSPC122, KLF12 zinc finger transcriptional repressor, Krueppel-like factor 12, Kruppel-like factor 12, Transcriptional repressor AP-2rep | |
Recombinant full-length human KLF12 protein (Uniprot: Q9Y4X4) | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
11278 | |
Store at 4°C in the dark. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction