Learn More
Invitrogen™ KLF12 Monoclonal Antibody (3D2)

Description
Peptide Sequence: MNIHMKRKTI KNINTFENRM LMLDGMPAVR VKTELLESEQ GSPNVHNYPD MEAVPLLLNN VKGEPPEDSL SVDHFQTQTE PVDLSINKAR.
Specifications
Specifications
| Antigen | KLF12 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3D2 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | KLF12 |
| Gene Accession No. | Q9Y4X4 |
| Gene Alias | 2700063E05Rik; AI225908; AI315654; AP-2 repressor; AP2REP; AP-2rep; AP-2rep transcription factor; B130052C06Rik; BHLHB24; D530033K05Rik; HSPC122; ID; KLF12; KLF12 zinc finger transcriptional repressor; Krueppel-like factor 12; Kruppel like factor 12; Kruppel-like factor 12; transcriptional repressor AP-2rep |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.