Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KLF12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584628
Description
Immunogen sequence: IKNINTFENR MLMLDGMPAV RVKTELLESE QGSPNVHNYP DMEAVPLLLN NVKGEPPEDS LSVDHFQTQT EPVDLSIN.
Activator protein-2 alpha (AP-2 alpha) is a developmentally-regulated transcription factor and important regulator of gene expression during vertebrate development and carcinogenesis. The protein encoded by this gene is a member of the Kruppel-like zinc finger protein family and can repress expression of the AP-2 alpha gene by binding to a specific site in the AP-2 alpha gene promoter. Repression by the encoded protein requires binding with a corepressor, CtBP1. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
KLF12 | |
Polyclonal | |
Unconjugated | |
KLF12 | |
2700063E05Rik; AI225908; AI315654; AP-2 repressor; AP2REP; AP-2rep; AP-2rep transcription factor; B130052C06Rik; BHLHB24; D530033K05Rik; HSPC122; ID; KLF12; KLF12 zinc finger transcriptional repressor; Krueppel-like factor 12; Kruppel like factor 12; Kruppel-like factor 12; transcriptional repressor AP-2rep | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11278 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y4X4 | |
KLF12 | |
Recombinant protein corresponding to Human KLF12. Recombinant protein control fragment (Product #RP-107313). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction