Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLF12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120729
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120729 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120729 Supplier Invitrogen™ Supplier No. PA5120729
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse brain, Rat brain. Immunogen sequence: GHGKAQMDPR GLSPRQSKSD SDDDDLPNVT LDSVNETGST ALSIARAVQE VHPSPVSRVR GNRMNNQKFP CSISPFSIES TRRQRRSESP D.

Activator protein-2 alpha (AP-2 alpha) is a developmentally-regulated transcription factor and important regulator of gene expression during vertebrate development and carcinogenesis. The protein encoded by this gene is a member of the Kruppel-like zinc finger protein family and can repress expression of the AP-2 alpha gene by binding to a specific site in the AP-2 alpha gene promoter. Repression by the encoded protein requires binding with a corepressor, CtBP1. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF12
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene KLF12
Gene Accession No. O35738
Gene Alias 2700063E05Rik; AI225908; AI315654; AP-2 repressor; AP2REP; AP-2rep; AP-2rep transcription factor; B130052C06Rik; BHLHB24; D530033K05Rik; HSPC122; ID; KLF12; KLF12 zinc finger transcriptional repressor; Krueppel-like factor 12; Kruppel like factor 12; Kruppel-like factor 12; transcriptional repressor AP-2rep
Gene Symbols KLF12
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 220-310 of human KLF12 (NP_009180.3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16597, 306110
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.