Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF13 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298809AF532
Description
KLF13 Polyclonal specifically detects KLF13 in Human samples. It is validated for Western Blot.Specifications
KLF13 | |
Polyclonal | |
Western Blot | |
basic transcription element binding protein 3, Basic transcription element-binding protein 3, BTEB3FKLF2, BTE-binding protein 3, Krueppel-like factor 13, Kruppel-like factor 13, Novel Sp1-like zinc finger transcription factor 1, NSLP1RFLAT1, RANTES factor of late activated T lymphocytes-1, RANTES factor of late activated T-lymphocytes 1, RFLAT-1FKLF-2, Sp1 like zinc finger transcription factor, Transcription factor BTEB3, Transcription factor NSLP1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human KLF13. | |
0.1 mL | |
Cancer, Chromatin Research, Transcription Factors and Regulators, Tumor Suppressors | |
51621 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction